Learn More
Aviva Systems Biology Corp RASSF2 Antibody - C-terminal region (ARP64234_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP64234_P050
This is a rabbit polyclonal antibody against RASSF2. It was validated on Western Blot. . Alternate names: DKFZp781O1747, KIAA0168, RASFADINApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-210