Learn More
Aviva Systems Biology Corp RASSF6 Antibody - middle region (ARP59685_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59685_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RASSF6. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human RASSF6. Alternate names: DKFZp686K23225ApplicationWB. Species reactivity: Human
Catalog No. 50-286-777