Learn More
Aviva Systems Biology Corp RASSF7 Antibody - middle region (ARP34389_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34389_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RASSF7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RASSF7. Alternate names: C11orf13, HRAS1, HRC1, MGC126069, MGC126070ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Yeast
Catalog No. 50-014-55373