Learn More
Aviva Systems Biology Corp RASSF8 Antibody - N-terminal region (ARP52301_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52301_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RASSF8. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human RASSF8. Alternate names: C12orf2, HoJ-1, HOJ1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-694