Learn More
Biomatik Corporation Rat 11-Beta-Hydroxysteroid Dehydrogenase Type 1 (HSD11b1) ELISA Kit
Supplier:  Biomatik Corporation EKU02003
Encompass_Preferred
Rat 11-Beta-Hydroxysteroid Dehydrogenase Type 1 (HSD11b1) ELISA Kit for detecting HSD11b1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P16232. Gene ID: 25116. Target Synonyms: 11DH; 11-beta-HSD1; HDL; HSD11; HSD11B; HSD11L; SDR26C1; Corticosteroid 11-beta-dehydrogenase isozyme 1; Short Chain Dehydrogenase/Reductase Family 26C,Member 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9160