Learn More
Biomatik Corporation Rat 17-Beta-Hydroxysteroid Dehydrogenase Type 3 (HSD17b3) ELISA Kit
Supplier:  Biomatik Corporation EKU10595
Encompass_Preferred
Rat 17-Beta-Hydroxysteroid Dehydrogenase Type 3 (HSD17b3) ELISA Kit for detecting HSD17b3 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O54939. Gene ID: 117182. Target Synonyms: SDR12C2; EDH17B3; Short Chain Dehydrogenase/Reductase Family 12C,Member 2; Testosterone 17-Beta-Dehydrogenase 3; Testicular 17-beta-hydroxysteroid dehydrogenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7415