Learn More
American Research Products Inc Rat 3 oXo 5 alpha steroid 4 de
Catalog No. 5000327675
Encompass
Rat 3-oXo-5-alpha-steroid 4-dehydrogenase 2 (SRD5A2) ELISA kit from CUSABIO Detection Range 6 pg/mL-1200 pg/mL Sensitivity 3 pg/mL Sample serum plasma tissue homogenates cell lysates Method Competitive
Catalog No. 50-003-27675 Supplier American Research Products Inc Supplier No. CSBEL022654RA
Rat 3-oXo-5-alpha-steroid 4-dehydrogenase 2 (SRD5A2) ELISA kit from CUSABIO Detection Range 6 pg/mL-1200 pg/mL Sensitivity 3 pg/mL Sample serum plasma tissue homogenates cell lysates Method Competitive