Learn More
ACROBiosystems ACROBiosystems Rat 4-1BB Ligand / TNFSF9 Protein, His,Flag Tag
Catalog No. 502106422
Encompass_Preferred
Recombinant Protein;100UG;Rat 4-1BB Ligand His,Flag Tag (41L-R52D3) is expressed from human 293 cells (HEK293). It contains AA Pro 106 - Gly 308 (Accession # Q80WE6-1).;This protein carries a polyhistidine tag at the N-terminus, followed by a flag tag. The protein has a calculated MW of 72.3 kDa.
Catalog No. 50-210-6422 Supplier ACROBiosystems Supplier No. 41LR52D3100UG
Recombinant Protein;100UG;Rat 4-1BB Ligand His,Flag Tag (41L-R52D3) is expressed from human 293 cells (HEK293). It contains AA Pro 106 - Gly 308 (Accession # Q80WE6-1).;This protein carries a polyhistidine tag at the N-terminus, followed by a flag tag. The protein has a calculated MW of 72.3 kDa.