Learn More
ACROBiosystems Rat 4-1BB / TNFRSF9 Protein, His Tag (MALS verified)
Supplier: ACROBiosystems 41BR52H3100UG
Recombinant Protein;100UG;Rat 4-1BB, His Tag (41B-R52H3) is expressed from human 293 cells (HEK293). It contains AA Thr 24 - Gln 187 (Accession # Q4W8J3-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.5 kDa. The protein migrates as 28-38 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.