Learn More
American Research Products Inc Rat 4 HydroXynonenal HNE ELI
Catalog No. 5000327990
Encompass
Rat 4-HydroXynonenal (HNE) ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma Method Competitive
Catalog No. 50-003-27990 Supplier American Research Products Inc Supplier No. CSBEQ027232RA
Rat 4-HydroXynonenal (HNE) ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma Method Competitive