Learn More
Biomatik Corporation Rat 4-HNE (4-Hydroxynonenal) ELISA Kit
Supplier:  Biomatik Corporation EKF58583
Encompass_Preferred
Rat 4-HNE (4-Hydroxynonenal) ELISA Kit for detecting 4-HNE in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: 4-HNE; 4-Hydroxynonenal; HNE; 4 HNE. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7793