Learn More
American Research Products Inc Rat 5 HydroXytryptamine 5HT E
Catalog No. 5000329708
Encompass
Rat 5-HydroXytryptamine 5HT ELISA Kit from CUSABIO Detection Range 0 8 ng/mL-160 ng/mL Sensitivity 0 4 ng/mL Sample serum plasma cell lysates tissue homogenates Method Competitive
Catalog No. 50-003-29708 Supplier American Research Products Inc Supplier No. CSBE08364R
Rat 5-HydroXytryptamine 5HT ELISA Kit from CUSABIO Detection Range 0 8 ng/mL-160 ng/mL Sensitivity 0 4 ng/mL Sample serum plasma cell lysates tissue homogenates Method Competitive