Learn More
American Research Products Inc Rat 5 nucleotidase cluster of
Catalog No. 5000329424
Encompass
Rat 5 -nucleotidase/cluster of differentiation 73 (NT5E/CD73) ELISA kit from CUSABIO Detection Range 0 625 mIU/mL-40 mIU/mL Sensitivity 0 156 mIU/mL Sample serum plasma tissue homogenates cell culture supernates Method Sandwich
Catalog No. 50-003-29424 Supplier American Research Products Inc Supplier No. CSBE14100R
Rat 5 -nucleotidase/cluster of differentiation 73 (NT5E/CD73) ELISA kit from CUSABIO Detection Range 0 625 mIU/mL-40 mIU/mL Sensitivity 0 156 mIU/mL Sample serum plasma tissue homogenates cell culture supernates Method Sandwich