Learn More
Biomatik Corporation Rat A Disintegrin And Metalloproteinase With Thrombospondin 5 (ADAMTS5) ELISA Kit
Supplier:  Biomatik Corporation EKU02060
Encompass_Preferred
Rat A Disintegrin And Metalloproteinase With Thrombospondin 5 (ADAMTS5) ELISA Kit for detecting ADAMTS5 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.116ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6TY19. Gene ID: 304135. Target Synonyms: ADMP-2; Aggrecanase-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5010