Learn More
Biomatik Corporation Rat A Disintegrin And Metalloproteinase With Thrombospondin 1 (ADAMTS1) ELISA Kit
Supplier:  Biomatik Corporation EKU02056
Encompass_Preferred
Rat A Disintegrin And Metalloproteinase With Thrombospondin 1 (ADAMTS1) ELISA Kit for detecting ADAMTS1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78.12-5000pg/ml. Sensitivity: 30.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9WUQ1. Target Synonyms: C3-C5; METH1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7127