Learn More
Biomatik Corporation Rat A Disintegrin And Metalloprotease 10 (ADAM10) CLIA Kit
Supplier:  Biomatik Corporation EKU09958
Encompass_Preferred
Rat A Disintegrin And Metalloprotease 10 (ADAM10) CLIA Kit for detecting ADAM10 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 6.86-5,000pg/ml. Sensitivity: 3.13pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q10743. Gene ID: 29650. Target Synonyms: CD156-C; CD156c; MADM; Kuz; CDw156; Kuzbanian protein homolog; Mammalian disintegrin-metalloprotease. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8394