Learn More
American Research Products Inc Rat a1 Acid glycoprotein a1 A
Catalog No. 5000329777
Encompass
Rat a1-Acid glycoprotein (a1-AGP) ELISA Kit from CUSABIO Detection Range 3 125 ng/mL-800 ng/mL Sensitivity 3 125 ng/mL Sample serum plasma urine cell culture supernates tissue homogenates Method Competitive
Catalog No. 50-003-29777 Supplier American Research Products Inc Supplier No. CSBE12730R
Rat a1-Acid glycoprotein (a1-AGP) ELISA Kit from CUSABIO Detection Range 3 125 ng/mL-800 ng/mL Sensitivity 3 125 ng/mL Sample serum plasma urine cell culture supernates tissue homogenates Method Competitive