Learn More
BIOHIPPO Rat a1AT Microsample
SDP
Catalog No. 5000314886
Encompass
Rat a1AT Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of a1AT in Serum plasma and other biological fluids It provides a detection range of 15 63-1000 g/mL with a sensitivity of 4 93 g/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat a1AT Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat a1AT Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat a1AT biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14886 Supplier BIOHIPPO Supplier No. BHE15212697
Rat a1AT Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of a1AT in Serum plasma and other biological fluids It provides a detection range of 15 63-1000 g/mL with a sensitivity of 4 93 g/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat a1AT Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat a1AT Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat a1AT biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color