Learn More
Sino Biological Rat ACE2 Matched ELISA Antibody Pair Set 15 Plates
Catalog No. 5000786653
Encompass_Preferred
The Rat ACE2 / Angiotensin-Converting Enzyme 2 ELISA Pair Set is for the quantitative determination of Rat ACE2 / Angiotensin-Converting Enzyme 2.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Rat ACE2 / Angiotensin-Converting Enzyme 2 coated on a 96-well plate. Standards and samples are added to the wells, and any Rat ACE2 / Angiotensin-Converting Enzyme 2 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Rat ACE2 / Angiotensin-Converting Enzyme 2 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Rat ACE2 / Angiotensin-Converting Enzyme 2 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm
Catalog No. 50-007-86653 Supplier Sino Biological Supplier No. SEK8003115
The Rat ACE2 / Angiotensin-Converting Enzyme 2 ELISA Pair Set is for the quantitative determination of Rat ACE2 / Angiotensin-Converting Enzyme 2.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Rat ACE2 / Angiotensin-Converting Enzyme 2 coated on a 96-well plate. Standards and samples are added to the wells, and any Rat ACE2 / Angiotensin-Converting Enzyme 2 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Rat ACE2 / Angiotensin-Converting Enzyme 2 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Rat ACE2 / Angiotensin-Converting Enzyme 2 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm