Learn More
Sino Biological Recombinant Rat ACE2 Protein (His Tag)
Supplier:  Sino Biological 80031R08H1
Encompass_Preferred
A DNA sequence encoding the rat ACE2 (Q5EGZ1) extracellular domain (Met 1-Thr 740) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-2857