Learn More
American Research Products Inc Rat Acetylcholinesterase AChE
Catalog No. 5000328968
Encompass
Rat Acetylcholinesterase AChE ELISA Kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28968 Supplier American Research Products Inc Supplier No. CSBE11304R
Rat Acetylcholinesterase AChE ELISA Kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich