Learn More
Innovative Research Inc Rat Act tPA Kit
Catalog No. 502743992
Encompass_Preferred
Rat Active tPA ELISA Kit - This rat tPA activity assay is intended for the quantitative determination of active tissue plasminogen activator (tPA) in rat plasma and other biological fluids. For research use only. Strip well format. Reagents for up to 96 tests. Functionally active rat tPA will form a covalent complex with the biotinylated human PAI-1 which is bound to the avidin on the plate. Only free active tPA will react with the PAI-1 bound to the plate. After appropriate washing steps, polyclonal anti-tPA primary antibody binds to the captured tPA. Excess antibody is washed away and bound primary antibody is then reacted with the secondary antibody conjugated to horseradish peroxidase. Following an additional washing step, TMB substrate is used for color development at 450nm. The amount of color development is directly proportional to the concentration of active tPA in the sample.
Catalog No. 50-274-3992 Supplier Innovative Research Inc Supplier No. IRTTPAKTA
Rat Active tPA ELISA Kit - This rat tPA activity assay is intended for the quantitative determination of active tissue plasminogen activator (tPA) in rat plasma and other biological fluids. For research use only. Strip well format. Reagents for up to 96 tests. Functionally active rat tPA will form a covalent complex with the biotinylated human PAI-1 which is bound to the avidin on the plate. Only free active tPA will react with the PAI-1 bound to the plate. After appropriate washing steps, polyclonal anti-tPA primary antibody binds to the captured tPA. Excess antibody is washed away and bound primary antibody is then reacted with the secondary antibody conjugated to horseradish peroxidase. Following an additional washing step, TMB substrate is used for color development at 450nm. The amount of color development is directly proportional to the concentration of active tPA in the sample.