Learn More
American Research Products Inc Rat actiVated coagulation fact
Catalog No. 5000327019
Encompass
Rat actiVated coagulation factor X (FXa) ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-27019 Supplier American Research Products Inc Supplier No. CSBEQ027251RA
Rat actiVated coagulation factor X (FXa) ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma tissue homogenates Method Competitive