Learn More
Biomatik Corporation Rat Activating Transcription Factor 6 (ATF6) ELISA Kit
Supplier:  Biomatik Corporation EKU12454
Encompass_Preferred
Rat Activating Transcription Factor 6 (ATF6) ELISA Kit for detecting ATF6 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: G3V909. Gene ID: 304962. Target Synonyms: Processed cyclic AMP-dependent transcription factor ATF-6 alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4690