Learn More
Biomatik Corporation Rat Activin A (ACVA) ELISA Kit
Supplier:  Biomatik Corporation EKU02112
Encompass_Preferred
Rat Activin A (ACVA) ELISA Kit for detecting ACVA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 4.71pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P18331. Gene ID: 29200. Target Synonyms: ACV-A; Activin Beta A Beta A Homodimer. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4945