Learn More
Biomatik Corporation Rat Activin B (ACVB) ELISA Kit
Supplier:  Biomatik Corporation EKU08668
Encompass_Preferred
Rat Activin B (ACVB) ELISA Kit for detecting ACVB in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 5.29pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P17491. Gene ID: 25196. Target Synonyms: ACV-B; Activin Beta B Beta B Homodimer. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7767