Learn More
Sino Biological Recombinant Rat ALK-1 Protein, HPLC-verified
Supplier:  Sino Biological 10596RCCH100
Encompass_Preferred
A DNA sequence encoding the Rat ACVRL1 (NP_071886.1) (Met1-Ala118) was expressed with six amino acids (ADDDDK) at the C-terminus.
Catalog No. 50-255-5250