Learn More
Stoelting™ Rat Adaptor Kit, 18 Degree for Parallel Rail Stereotaxic, Series 51800 and 51850
SDP
Catalog No. 10000218 Shop All Stoelting Co Products
Change view
Click to view available options
For Use With (Application):
Neuroscience
Catalog No. For Use With (Application)
10-000-218 Neuroscience
1 options
Catalog No. 10-000-218 Supplier Stoelting™ Supplier No. 51821
Add to Cart
Add to Cart
Nose clamp assembly and ear bars are included.
  • 30 mm of vertical travel along dovetail slide and 0.1 mm vernier scales
  • 50 mm of Anterior-Posterior travel possible
Specifications
For Use With (Application) Neuroscience