Learn More
World Precision Instrument Rat Adaptor Kit, 45 Deg
SDP
Catalog No. 502539177
Encompass
The 505239Rat Adaptor Kit, 18 Degree is included with all WPI Rat Stereotaxic Instruments. It has 30 mm of vertical travel along dovetail slide and 0.1 mm vernier scales. 50 mm of Anterior-Posterior travel possible. The nose clamp assembly, and either standard 18 deg. (traditional) Ear Bars or non-puncture 45° Ear Bars, are included. For nontraumatic ear bars select the 45° option (505238).
Catalog No. 50-253-9177 Supplier World Precision Instrument Supplier No. 505238
The 505239Rat Adaptor Kit, 18 Degree is included with all WPI Rat Stereotaxic Instruments. It has 30 mm of vertical travel along dovetail slide and 0.1 mm vernier scales. 50 mm of Anterior-Posterior travel possible. The nose clamp assembly, and either standard 18 deg. (traditional) Ear Bars or non-puncture 45° Ear Bars, are included. For nontraumatic ear bars select the 45° option (505238).