Learn More
Biomatik Corporation Rat Adenylyl Cyclase Associated Protein 1 (CAP1) ELISA Kit
Supplier:  Biomatik Corporation EKU11958
Encompass_Preferred
Rat Adenylyl Cyclase Associated Protein 1 (CAP1) ELISA Kit for detecting CAP1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q08163. Gene ID: 64185. Target Synonyms: CAP; CAP1-PEN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8059