Learn More
Raybiotech Inc Rat Adiponectin (ACRP30) ELISA Kit
Supplier:  Raybiotech Inc ELRADIPONECTIN1
Encompass_Preferred
For Cell Culture Supernatants and Serum samples. Sensitivity: 0.8 ng/ml . Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Rat Adiponectin (ADIPOQ/ACRP30)
Catalog No. 50-195-0213