Learn More
Crystal Chem Inc Rat Adiponectin ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80570
Encompass
The Rat Adiponectin ELISA is used to quantify levels of rat adiponectin in serum and plasma using only a 5 µL sample volume. Highlights: Kits use only 5 µL sample, ideal for rat applications. Dynamic range of 0.25 - 10 ng/mL. High sensitivity of 0.08 ng/mL. Results in under 2.5 hours.Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7737