Learn More
Biomatik Corporation Rat Adiponectin Receptor 2 (ADIPOR2) ELISA Kit
Supplier:  Biomatik Corporation EKU02162
Encompass_Preferred
Rat Adiponectin Receptor 2 (ADIPOR2) ELISA Kit for detecting ADIPOR2 in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.122ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q3YAF7. Gene ID: 312670. Target Synonyms: PAQR2; ACDCR2; Progestin and AdipoQ Receptor Family Member II. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7311