Learn More
Biomatik Corporation Rat Adipose Differentiation Related Protein (ADRP) ELISA Kit
Supplier:  Biomatik Corporation EKU09059
Encompass_Preferred
Rat Adipose Differentiation Related Protein (ADRP) ELISA Kit for detecting ADRP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.57ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q5U2U5. Gene ID: 298199. Target Synonyms: PLIN2; ADFP; Adipophilin; Perilipin 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8572