Learn More
Biomatik Corporation Rat Adrenocorticotropic Hormone (ACTH) CLIA Kit
Supplier:  Biomatik Corporation EKU08643
Encompass_Preferred
Rat Adrenocorticotropic Hormone (ACTH) CLIA Kit for detecting ACTH in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 11.72-3000pg/ml. Sensitivity: 4.52pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Accession Number: P01194. Target Synonyms: Corticotropin; Adrenocorticotropin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6853