Learn More
American Research Products Inc Rat adrenocorticotropic hormon
Catalog No. 5000329631
Encompass
Rat adrenocorticotropic hormone (ACTH) ELISA kit from CUSABIO Detection Range 1 25 pg/mL-50 pg/mL Sensitivity 1 25 pg/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-29631 Supplier American Research Products Inc Supplier No. CSBE06875R
Rat adrenocorticotropic hormone (ACTH) ELISA kit from CUSABIO Detection Range 1 25 pg/mL-50 pg/mL Sensitivity 1 25 pg/mL Sample serum plasma tissue homogenates Method Competitive