Learn More
American Research Products Inc Rat adVanced glycation end pro
Catalog No. 5000330137
Encompass
Rat adVanced glycation end products AGEs ELISA Kit from CUSABIO Detection Range 3 9 g/mL-250 g/mL Sensitivity 0 97 g/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-30137 Supplier American Research Products Inc Supplier No. CSBE09413R
Rat adVanced glycation end products AGEs ELISA Kit from CUSABIO Detection Range 3 9 g/mL-250 g/mL Sensitivity 0 97 g/mL Sample serum plasma tissue homogenates Method Sandwich