Learn More
American Research Products Inc Rat AdVanced glycosylation end
Catalog No. 5000328537
Encompass
Rat AdVanced glycosylation end product-specific receptor (AGER) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28537 Supplier American Research Products Inc Supplier No. CSBEL001441RA
Rat AdVanced glycosylation end product-specific receptor (AGER) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates Method Sandwich