Learn More
American Research Products Inc Rat adVanced oXidation protein
Catalog No. 5000328290
Encompass
Rat adVanced oXidation protein products (AOPP) ELISA kit from CUSABIO Detection Range 15 6 nmol/mL-1000 nmol/mL Sensitivity 3 9 nmol/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28290 Supplier American Research Products Inc Supplier No. CSBEQ027429RA
Rat adVanced oXidation protein products (AOPP) ELISA kit from CUSABIO Detection Range 15 6 nmol/mL-1000 nmol/mL Sensitivity 3 9 nmol/mL Sample serum plasma tissue homogenates Method Sandwich