Learn More
Biomatik Corporation Rat Afamin (AFM) ELISA Kit
Supplier:  Biomatik Corporation EKU02199
Encompass_Preferred
Rat Afamin (AFM) ELISA Kit for detecting AFM in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 2.47-200ng/ml. Sensitivity: 0.93ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P36953. Gene ID: 282708. Target Synonyms: ALB2; ALBA; ALF; A-ALB; Alpha-Albumin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9147