Learn More
Biomatik Corporation Rat Agouti Related Protein (AGRP) ELISA Kit
Supplier:  Biomatik Corporation EKU02206
Encompass_Preferred
Rat Agouti Related Protein (AGRP) ELISA Kit for detecting AGRP in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 9.4-600pg/ml. Sensitivity: 4.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: F1MAG1. Gene ID: 25582. Target Synonyms: AGRT; ASIP2; ART; Agouti-Related Peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9146