Learn More
Crystal Chem Inc Rat Alpha Glutathione S-Transferase (AGST) ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80675
Encompass
The Rat Glutathione S-Transferase Alpha (AGST) ELISA is a highly sensitive assay that is used to quantify levels of in serum, plasma, and urine. Highlights: Kit uses sample size of only 5 µL. High sensitivity of 1.5 ng/mL. Dynamic range of 3.125 - 100 ng/mL. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7802