Learn More
Sino Biological Anti-Adenylate Kinase 1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80583T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat AK1 / Adenylate kinase 1 (rR AK1 / Adenylate kinase 1; Catalog#80583-R07E; NP_077325.2; Met1-Lys194). AK1 / Adenylate kinase 1 specific IgG was purified by Rat AK1 / Adenylate kinase 1 affinity chromatography.
Catalog No. 50-255-2767