Learn More
Raybiotech Inc C-Series Rat Acute Kidney Injury Antibody Array 1 Kit.
Catalog No. 501948764
Encompass_Preferred
Detects 7 rat Cytokines. Suitable for all liquid sample types. 4 sample kit. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: TIM-1 (KIM-1), FABP1, MCP-1 (CCL2), Cystatin C, Lipocalin-2 (NGAL), TIMP-1, VEGF-A
Catalog No. 50-194-8764 Supplier Raybiotech Inc Supplier No. AARAKI12
Detects 7 rat Cytokines. Suitable for all liquid sample types. 4 sample kit. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: TIM-1 (KIM-1), FABP1, MCP-1 (CCL2), Cystatin C, Lipocalin-2 (NGAL), TIMP-1, VEGF-A