Learn More
Raybiotech Inc G-Series Rat Acute Kidney Injury Antibody Array 1 Kit.
Catalog No. 501948767
Encompass_Preferred
Detects 7 rat Cytokines. Suitable for all liquid sample types. 4 sample kit. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: TIM-1 (KIM-1), FABP1, MCP-1 (CCL2), Cystatin C, Lipocalin-2 (NGAL), TIMP-1, VEGF-A
Catalog No. 50-194-8767 Supplier Raybiotech Inc Supplier No. AARAKIG14
Detects 7 rat Cytokines. Suitable for all liquid sample types. 4 sample kit. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: TIM-1 (KIM-1), FABP1, MCP-1 (CCL2), Cystatin C, Lipocalin-2 (NGAL), TIMP-1, VEGF-A