Learn More
American Research Products Inc Rat Alanine aminotransferase
Catalog No. 5000329042
Encompass
Rat Alanine aminotransferase (ALT) ELISA Kit from CUSABIO Detection Range 3 12 mIU/mL-200 mIU/mL Sensitivity 0 78 mIU/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-29042 Supplier American Research Products Inc Supplier No. CSBE13024R
Rat Alanine aminotransferase (ALT) ELISA Kit from CUSABIO Detection Range 3 12 mIU/mL-200 mIU/mL Sensitivity 0 78 mIU/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich