Learn More
Biomatik Corporation Rat Alanine Aminotransferase (ALT) CLIA Kit
Supplier:  Biomatik Corporation EKU09786
Encompass_Preferred
Rat Alanine Aminotransferase (ALT) CLIA Kit for detecting ALT in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.14-100ng/ml. Sensitivity: 0.05ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P25409. Gene ID: 81670. Target Synonyms: GPT; ALAT; SGPT; AAT1; GPT1; Serum Glutamic Pyruvic Transaminase; Alanine Transaminase; Glutamate pyruvate transaminase 1; Glutamic--alanine transaminase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8435