Learn More
Biomatik Corporation Rat Albumin (ALB) CLIA Kit
Supplier:  Biomatik Corporation EKU08655
Encompass_Preferred
Rat Albumin (ALB) CLIA Kit for detecting ALB in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 6.17-500ug/ml. Sensitivity: 2.17ug/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Accession Number: P02770. Gene ID: 24186. Target Synonyms: Serum albumin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5142