Learn More
Bon Opus Biosciences Recombinant Rat Albumin (C-His) 50ug
SDP
Supplier:  Bon Opus Biosciences BP110150UG
Encompass_Preferred
Recombinant Rat Albumin (C-His) is produced by Human Cells. The target gene encoding Glu25-Ala608 is expressed with a C-His tag. Accession# P02770. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1101
Catalog No. 50-217-2698