Learn More
American Research Products Inc Rat aldehyde dehydrogenase AL
Catalog No. 5000326207
Encompass
Rat aldehyde dehydrogenase ALDH ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26207 Supplier American Research Products Inc Supplier No. CSBE11179R
Rat aldehyde dehydrogenase ALDH ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich